Learn More
Medchemexpress LLC Heparin | 9005-49-6 | >98.0% | 10 MG
SDP
Catalog No. 5000298788
Encompass_Preferred
Heparin is a highly sulfated glycosaminoglycan used as an anticoagulant reagent in biochemical and cellular research. This unfractionated form is supplied as a purified powder suitable for in vitro assays, anticoagulation controls, and studies of coagulation-related processes.
  • High purity >98%.
  • Polydisperse glycosaminoglycan; molecular weight varies between preparations.
  • Suitable for in vitro anticoagulant assays and biochemical studies.
  • Available in multiple pack sizes for small-scale research use.
  • Available in sodium and lithium salt forms for different experimental requirements.
Catalog No. 50-002-98788 Supplier Medchemexpress LLC Supplier No. HY1756710MG
Heparin is a highly sulfated glycosaminoglycan used as an anticoagulant reagent in biochemical and cellular research. This unfractionated form is supplied as a purified powder suitable for in vitro assays, anticoagulation controls, and studies of coagulation-related processes.
  • High purity >98%.
  • Polydisperse glycosaminoglycan; molecular weight varies between preparations.
  • Suitable for in vitro anticoagulant assays and biochemical studies.
  • Available in multiple pack sizes for small-scale research use.
  • Available in sodium and lithium salt forms for different experimental requirements.