Learn More
Cayman Chemical HeparIn-bIndng PeptIdetrIf 1mg
SDP
Catalog No. 502914754
Encompass_Preferred
A peptide fragment of fibronectin; induces focal adhesion formation in pre-spread HEFs from 1-1,000 UG/ml; inhibits HBFN-f-induced production of MMP-1, MMP-2, and MMP-9 in isolated human articular cartilage; has been used in peptide surface micropatterning for surface endothelialization
Catalog No. 50-291-4754 Supplier Cayman Chemical Supplier No. 397251 MG
A peptide fragment of fibronectin; induces focal adhesion formation in pre-spread HEFs from 1-1,000 UG/ml; inhibits HBFN-f-induced production of MMP-1, MMP-2, and MMP-9 in isolated human articular cartilage; has been used in peptide surface micropatterning for surface endothelialization