Learn More
Medchemexpress LLC Heparin calcium | 37270-89-6 | 3600-5000 Da | 50 MG
SDP
Catalog No. 5000242238
Encompass_Preferred
Heparin calcium (MW 3600-5000) is a low-molecular-weight heparin calcium salt used as an anticoagulant reagent for biochemical and pharmacological research. It forms a complex with antithrombin III to inhibit thrombin and other activated clotting factors, preventing fibrin formation. The material is a heterogeneous sulfated polysaccharide mixture provided as a dry reagent for laboratory use.
  • Molecular weight 3600-5000 Da
  • Anticoagulant activity via antithrombin III-mediated inhibition of clotting factors
  • Intended for biochemical and pharmacological research applications
  • Heterogeneous sulfated polysaccharide mixture, not a single defined molecule
  • Supplied as a dry reagent suitable for in vitro assays
Catalog No. 50-002-42238 Supplier Medchemexpress LLC Supplier No. HY107966A50MG
Heparin calcium (MW 3600-5000) is a low-molecular-weight heparin calcium salt used as an anticoagulant reagent for biochemical and pharmacological research. It forms a complex with antithrombin III to inhibit thrombin and other activated clotting factors, preventing fibrin formation. The material is a heterogeneous sulfated polysaccharide mixture provided as a dry reagent for laboratory use.
  • Molecular weight 3600-5000 Da
  • Anticoagulant activity via antithrombin III-mediated inhibition of clotting factors
  • Intended for biochemical and pharmacological research applications
  • Heterogeneous sulfated polysaccharide mixture, not a single defined molecule
  • Supplied as a dry reagent suitable for in vitro assays