Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Heparin calcium | 37270-89-6 | MFCD00131296 | ≥98.0% | 3600-5000 | Not specified (heterogeneous polysaccharide; no single chemical formula) | 25 MG
SDP

Catalog No. 5000241981
Encompass_Preferred

Heparin calcium (MW 3600-5000) is a low-molecular-weight heparin anticoagulant supplied for research use. It binds antithrombin III to inactivate thrombin and other activated clotting factors, preventing fibrin formation. The material is provided as a white to off-white solid with good aqueous solubility and is intended for laboratory assays involving anticoagulation or coagulation pathway studies.

  • Intended for research use only.
  • Anticoagulant that binds antithrombin III to inhibit coagulation.
  • Molecular weight range 3600-5000.
  • Supplied as a white to off-white solid.
  • Soluble in water (≈200 mg/mL with ultrasonication).
  • Store sealed and dry at 4 °C; in solution, store at -20 °C to -80 °C per guidelines.
  • Typical purity ≥98% (batch-specific certificate of analysis available).
  • Available in small laboratory package sizes (example: 25 mg).

Catalog No. 50-002-41981 Supplier Medchemexpress LLC Supplier No. HY107966A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Heparin calcium (MW 3600-5000) is a low-molecular-weight heparin anticoagulant supplied for research use. It binds antithrombin III to inactivate thrombin and other activated clotting factors, preventing fibrin formation. The material is provided as a white to off-white solid with good aqueous solubility and is intended for laboratory assays involving anticoagulation or coagulation pathway studies.

  • Intended for research use only.
  • Anticoagulant that binds antithrombin III to inhibit coagulation.
  • Molecular weight range 3600-5000.
  • Supplied as a white to off-white solid.
  • Soluble in water (≈200 mg/mL with ultrasonication).
  • Store sealed and dry at 4 °C; in solution, store at -20 °C to -80 °C per guidelines.
  • Typical purity ≥98% (batch-specific certificate of analysis available).
  • Available in small laboratory package sizes (example: 25 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.