Learn More
Medchemexpress LLC Heparin calcium (MW 3600-5000) | 37270-89-6 | 3600-5000 Da | 100 MG
SDP
Catalog No. 5000241793
Encompass_Preferred
Heparin calcium (MW 3600-5000) is the calcium salt form of heparin, a polydisperse sulfated glycosaminoglycan anticoagulant supplied as a solid for research use. It acts by binding antithrombin III, accelerating inactivation of thrombin and other activated clotting factors to prevent fibrin formation. Supplied in low-molecular-weight fractions with an average molecular weight range of 3600-5000 Da.
  • Anticoagulant action via antithrombin III binding.
  • Low molecular weight range: 3600-5000 Da.
  • Calcium salt supplied as a solid form.
  • Intended for research use only, not for clinical use.
  • Common pack size: 100 mg.
Catalog No. 50-002-41793 Supplier Medchemexpress LLC Supplier No. HY107966A100MG
Heparin calcium (MW 3600-5000) is the calcium salt form of heparin, a polydisperse sulfated glycosaminoglycan anticoagulant supplied as a solid for research use. It acts by binding antithrombin III, accelerating inactivation of thrombin and other activated clotting factors to prevent fibrin formation. Supplied in low-molecular-weight fractions with an average molecular weight range of 3600-5000 Da.
  • Anticoagulant action via antithrombin III binding.
  • Low molecular weight range: 3600-5000 Da.
  • Calcium salt supplied as a solid form.
  • Intended for research use only, not for clinical use.
  • Common pack size: 100 mg.