Learn More
Med Vet International Heparin Coated Microhematocrit Tubes, 75mm, J0543M, 200 Pack
Catalog No. 5000582330
Encompass_Preferred
Wrapped with three layers of Mylar. Hemato-clad hematocrit tubes combine the precision of glass with the safety provided by a mylar overwrap. Accidental breakage is significantly reduced particularly when pressing into sealing clay. In the event of breakage, the Mylar wrapping will help contain the blood sample and provides an added layer of protection against cuts from broken glass. Handy dispensing vial with chrome lid.
Catalog No. 50-005-82330 Supplier Med Vet International Supplier No. JOR543M
Wrapped with three layers of Mylar. Hemato-clad hematocrit tubes combine the precision of glass with the safety provided by a mylar overwrap. Accidental breakage is significantly reduced particularly when pressing into sealing clay. In the event of breakage, the Mylar wrapping will help contain the blood sample and provides an added layer of protection against cuts from broken glass. Handy dispensing vial with chrome lid.