Learn More
Bioworld Heparin-Coated Microplates (Clear), 25 Plate(s)
Catalog No. 50197532
Encompass
High quality microplates coated with a high binding heparin solution.Heparin, Sodium salt is a low molecular weight heparin (LMWH) polymer that produces its major anticoagulant effect by activating antithrombin. This activation causes a conformational change in ATIII and allows for increased flexibility in its reactive site loop. Heparin is a highly sulfated glycosaminoglycan known for preventing clots. Heparin, Sodium Salt is also an activator of RyR and ATIII.
Catalog No. 50-197-532 Supplier Bioworld Supplier No. 201400054
High quality microplates coated with a high binding heparin solution.Heparin, Sodium salt is a low molecular weight heparin (LMWH) polymer that produces its major anticoagulant effect by activating antithrombin. This activation causes a conformational change in ATIII and allows for increased flexibility in its reactive site loop. Heparin is a highly sulfated glycosaminoglycan known for preventing clots. Heparin, Sodium Salt is also an activator of RyR and ATIII.