Learn More
Bioworld Heparin-Coated Microplates (Clear) , 10 plates
Catalog No. 50197531
Encompass
High quality microplates coated with a high binding heparin solution. Heparin, Sodium salt is a low molecular weight heparin (LMWH) polymer that produces its major anticoagulant effect by activating antithrombin. This activation causes a conformational change in ATIII and allows for increased flexibility in its reactive site loop. Heparin is a highly sulfated glycosaminoglycan known for preventing clots. Heparin, Sodium Salt is also an activator of RyR and ATIII. 10 plates
Catalog No. 50-197-531 Supplier Bioworld Supplier No. 201400053
High quality microplates coated with a high binding heparin solution. Heparin, Sodium salt is a low molecular weight heparin (LMWH) polymer that produces its major anticoagulant effect by activating antithrombin. This activation causes a conformational change in ATIII and allows for increased flexibility in its reactive site loop. Heparin is a highly sulfated glycosaminoglycan known for preventing clots. Heparin, Sodium Salt is also an activator of RyR and ATIII. 10 plates