Learn More
Haematologic Technologies Inc Human Heparin Cofactor II, 100 ug
Supplier:  Haematologic Technologies Inc HCII0190
Encompass_Preferred
Heparin cofactor II (HCII) (heparin cofactor A, antithrombin BM, dermatan sulfate cofactor, human leuserpin-2) is a single chain glycoprotein (Mr=65,600, 10% carbohydrate) member of the serine protease inhibitor (serpin) family. Amino acid sequence analysis of heparin cofactor II reveals 3 potential N-glycosylation sites and 2 sulfated tyrosines located in two internal repeats of 7 residues located near the NH2-terminal. The plasma concentration is estimated to be 90 ug/ml.
Catalog No. 50-883-421