Learn More
Apexbio Technology LLC heparin cofactor II precursor (SERPIND1) fragment [Homo sapiens] 5mg
SDP
Supplier:  Apexbio Technology LLC A11435
Encompass_Preferred
Heparin cofactor II precursor SERPIND1 fragment is a peptide fragment targeting thrombin (factor IIa) It is designed to regulate thrombin activity thereby modulating blood coagulation pathways Heparin cofactor II precursor SERPIND1 fragment exerts its biological effect through inhibition of thrombin with its activity accelerated by glycosaminoglycans such as heparin or dermatan sulfate Based on these properties heparin cofactor II precursor SERPIND1 fragment has potential applications in investigating thrombin regulation and in studies of serpin-protease interactions within biomedical research
Catalog No. 50-000-96934