Learn More
Medchemexpress LLC Heparin disaccharide I-S tetrasodium | 136098-10-7 | MFCD00466927 | 99.0% | C12H15NNa4O19S3 | 10MG
SDP
Catalog No. 5000258921
Encompass_Preferred
Heparin disaccharide I-S tetrasodium is a purified heparin-derived disaccharide supplied as the tetrasodium salt for biochemical assays and glycosaminoglycan research. It is the major disaccharide produced by heparinases I and II from heparin and is provided as an analytical-grade reagent.
  • Purified tetrasodium form suitable for analytical assays.
  • Major disaccharide produced by heparinases I and II.
  • High purity: 99.0%.
  • Molecular weight: 665.40 g/mol.
  • Available in small research pack sizes (1 mg-100 mg).
  • Suitable for biochemical assay and glycosaminoglycan research.
Catalog No. 50-002-58921 Supplier Medchemexpress LLC Supplier No. HY14162710MG
Heparin disaccharide I-S tetrasodium is a purified heparin-derived disaccharide supplied as the tetrasodium salt for biochemical assays and glycosaminoglycan research. It is the major disaccharide produced by heparinases I and II from heparin and is provided as an analytical-grade reagent.
  • Purified tetrasodium form suitable for analytical assays.
  • Major disaccharide produced by heparinases I and II.
  • High purity: 99.0%.
  • Molecular weight: 665.40 g/mol.
  • Available in small research pack sizes (1 mg-100 mg).
  • Suitable for biochemical assay and glycosaminoglycan research.