Learn More
Braintree Scientific Heparin/Glycerol Catheter Loc Solution 10ml vial, 5/pk
SDP
Catalog No. 501954934
Encompass
Cath-Loc HGS: Heparin/Glycerol Catheter Lock Solution. The antithrombic characteristics of heparin plus the viscous nature of glycerol is combined in a solution to promote and extend the patency of vascular catheters. Proven as the solution of choice in studies in catheterized animals*, Cath-Loc HGS solution acts as a non-toxic anticoagulant lock solution for indwelling catheters. This formulation is the most widely used as a pre-clinical catheter lock. *Y.S. Luo, et. al. AALAS 2000 Comparison of Catheter Lock Solutions in Rats. Not for human use - animal research use only. *An Accountability form, completed by a licensed professional, will be required to purchase this item. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4934 Supplier Braintree Scientific Supplier No. HGS5
Cath-Loc HGS: Heparin/Glycerol Catheter Lock Solution. The antithrombic characteristics of heparin plus the viscous nature of glycerol is combined in a solution to promote and extend the patency of vascular catheters. Proven as the solution of choice in studies in catheterized animals*, Cath-Loc HGS solution acts as a non-toxic anticoagulant lock solution for indwelling catheters. This formulation is the most widely used as a pre-clinical catheter lock. *Y.S. Luo, et. al. AALAS 2000 Comparison of Catheter Lock Solutions in Rats. Not for human use - animal research use only. *An Accountability form, completed by a licensed professional, will be required to purchase this item. **Sold for research purposes only and not for human use**