Learn More
Apexbio Technology LLC HyperTrap Heparin HP Column(Synonyms: Heparin Affinity Column, Heparin HP Column, HyperTrap Heparin Affinity Chromatography Column), 5x1ml.
SDP
Catalog No. 5000061134
Encompass_Preferred
HyperTrap Heparin HP Column features the HyperChrom Heparin HP Agarose affinity chromatography resin which consists of heparin covalently immobilized on an agarose matrix Heparin is a naturally occurring glycosaminoglycan known for specific affinity interactions with various biomolecules including coagulation factors growth factors interferons lipoprotein lipase nucleic acid-binding proteins steroid receptors and antithrombin III This column enables efficient separation and purification of these proteins making it suitable for biochemical research proteomics analyses and protein characterization studies
Catalog No. 50-000-61134 Supplier Apexbio Technology LLC Supplier No. PC10095
HyperTrap Heparin HP Column features the HyperChrom Heparin HP Agarose affinity chromatography resin which consists of heparin covalently immobilized on an agarose matrix Heparin is a naturally occurring glycosaminoglycan known for specific affinity interactions with various biomolecules including coagulation factors growth factors interferons lipoprotein lipase nucleic acid-binding proteins steroid receptors and antithrombin III This column enables efficient separation and purification of these proteins making it suitable for biochemical research proteomics analyses and protein characterization studies