Learn More
Sigma Aldrich Fine Chemicals Biosciences Heparin lithium salt from porcine intestinal mucosa, 9045-22-1, MFCD00131308, 100 KU
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H0878100KU
Encompass
≥150 USP units/mg. Heparin is a microheterogeneous molecule widely used and studied as an anticoagulation drug.
Catalog No. NC1074539