Learn More
Medchemexpress LLC HY-17567B 100mg Medchemexpress, Heparin (Lithium salt) CAS:9045-22-1 Purity:>98%
SDP
Catalog No. 501969995
Encompass_Preferred
Medchemexpress, HY-17567B 100mg Heparin (Lithium salt) CAS:9045-22-1 Heparin Lithium salt is an anticoagulant which binds reversibly to antithrombin III (ATIII). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9995 Supplier Medchemexpress LLC Supplier No. HY17567B100MG
Medchemexpress, HY-17567B 100mg Heparin (Lithium salt) CAS:9045-22-1 Heparin Lithium salt is an anticoagulant which binds reversibly to antithrombin III (ATIII). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.