Learn More
Medchemexpress LLC Heparin Lithium salt | 9045-22-1 | 500 MG
SDP
Catalog No. 5000387377
Encompass_Preferred
Heparin Lithium salt is an anticoagulant that binds reversibly to antithrombin III (ATIII) and significantly inhibits exosome-cell interactions.
  • Anticoagulant
  • Binds reversibly to antithrombin III (ATIII)
  • Significantly inhibits exosome-cell interactions
Catalog No. 50-003-87377 Supplier Medchemexpress LLC Supplier No. HY17567B500MG
Heparin Lithium salt is an anticoagulant that binds reversibly to antithrombin III (ATIII) and significantly inhibits exosome-cell interactions.
  • Anticoagulant
  • Binds reversibly to antithrombin III (ATIII)
  • Significantly inhibits exosome-cell interactions