Learn More
Medchemexpress LLC Heparin Lithium salt | 9045-22-1 | MFCD00131308 | 1 G
SDP
Catalog No. 5000387156
Encompass_Preferred
Heparin Lithium salt is an anticoagulant that reversibly binds to antithrombin III (ATIII). It significantly inhibits exosome-cell interactions. Heparin is a potent anticoagulant drug, accelerating the rate at which antithrombin inhibits serine proteases in the blood coagulation cascade.
  • Anticoagulant
  • Reversibly binds to antithrombin III (ATIII)
  • Inhibits exosome-cell interactions
  • Enhances AT-III-mediated inhibition of blood coagulation factors
  • Promotes FGFR dimerization
  • For research use only
Catalog No. 50-003-87156 Supplier Medchemexpress LLC Supplier No. HY175671G
Heparin Lithium salt is an anticoagulant that reversibly binds to antithrombin III (ATIII). It significantly inhibits exosome-cell interactions. Heparin is a potent anticoagulant drug, accelerating the rate at which antithrombin inhibits serine proteases in the blood coagulation cascade.
  • Anticoagulant
  • Reversibly binds to antithrombin III (ATIII)
  • Inhibits exosome-cell interactions
  • Enhances AT-III-mediated inhibition of blood coagulation factors
  • Promotes FGFR dimerization
  • For research use only