Learn More
Medchemexpress LLC Heparin methacrylate | 10 MG
SDP
Catalog No. 5000288121
Encompass_Preferred
Heparin methacrylate (HepMA) is a methacrylated heparin polymer used as a tissue engineering scaffold and 3D printing bioink. It crosslinks under photoinitiation to form fibrous hydrogels that present bioactive adhesion sites, support cell attachment and growth, and enable biodegradable scaffolds for cell culture and tissue engineering research.
  • Methacrylated heparin polymer suitable for scaffold and bioink applications.
  • Crosslinks with photoinitiators to form stable hydrogels.
  • Presents bioactive adhesion sites to support cell interaction.
  • White to off-white solid formulation for lab handling.
  • Available in small research packages (e.g., 10 mg) with COA and SDS documentation.
  • Store as powder at recommended cold temperatures to preserve stability.
Catalog No. 50-002-88121 Supplier Medchemexpress LLC Supplier No. HY15822910MG
Heparin methacrylate (HepMA) is a methacrylated heparin polymer used as a tissue engineering scaffold and 3D printing bioink. It crosslinks under photoinitiation to form fibrous hydrogels that present bioactive adhesion sites, support cell attachment and growth, and enable biodegradable scaffolds for cell culture and tissue engineering research.
  • Methacrylated heparin polymer suitable for scaffold and bioink applications.
  • Crosslinks with photoinitiators to form stable hydrogels.
  • Presents bioactive adhesion sites to support cell interaction.
  • White to off-white solid formulation for lab handling.
  • Available in small research packages (e.g., 10 mg) with COA and SDS documentation.
  • Store as powder at recommended cold temperatures to preserve stability.