Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Heparin, Porcine Intestinal Mucosa, Biotin Conjugate, Calbiochem™,

Catalog No. 37505410MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-505-410MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-505-410MG Supplier MilliporeSigma™ Supplier No. 37505410MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 9005-49-6
Molecular Formula C26H42N2O37S5
Molecular Weight (g/mol) 1134.9
InChI Key HTTJABKRGRZYRN-UHFFFAOYNA-N
Synonym heparin, enoxaparin, clexane, lovenox, ardeparin, lmwh, lovenox hp, low molecular weight heparin, semuloparin, lipo-hepin
PubChem CID 772

Specifications

Packaging Glass Bottle
Physical Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.