Learn More
Sigma Aldrich Fine Chemicals Biosciences Heparin SepharoseR 6 Fast
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17099801
Encompass
Heparin Sepharose(R) 6 Fast Flow is a well established BioProcess affinity medium designed for capture or intermediate purification steps. This medium is extensively used for industrial purification of several marketed plasma proteins.Heparin Sepharose(R) 6 Fast Flow is a member of the Cytiva range of affinity chromatography media for capture and intermediate purification.Heparin Sepharose(R) 6 Fast Flow is composed of cross-linked 6% agarose beads modified sodium heparin. Immobilization is performed using chemically stable and oriented coupling giving high binding capacity. The chemical stability of the media is solely dictated by the heparin ligand.This medium allows for fast and reliable separations of biomolecules with an affinity for heparin including antithrombin III coagulation factors and other plasma proteins. Additional applications include DNA binding proteins lipoproteins protein synthesis factors enzymes that act on nucleic acids and steroid receptors.Heparin Sepharo
Catalog No. 50-290-7285