Learn More
SAI Infusion Technologies Heparin Sodium - 30ml vials
SDP
Catalog No. NC3306829
Encompass
Heparin Sodium Injection, USP. 30,000USP units per 30ml vial, 1,000USP units per ml. Not for lock flush. 25 vials/case. Product is for laboratory animal research only.
Catalog No. NC3306829 Supplier SAI Infusion Technologies Supplier No. HS10030C
Heparin Sodium Injection, USP. 30,000USP units per 30ml vial, 1,000USP units per ml. Not for lock flush. 25 vials/case. Product is for laboratory animal research only.