Learn More
Apexbio Technology LLC Heparin sodium, 100mg Cas# 9041-08-1
SDP
Catalog No. 501998522
Encompass_Preferred
Minimum activity: > 150 I.U./mg. Glycosaminoglycan that behaves as an anticoagulant. Binds with high affinity to antithrombin III (AT-III). Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8522 Supplier Apexbio Technology LLC Supplier No. A5066100
Minimum activity: > 150 I.U./mg. Glycosaminoglycan that behaves as an anticoagulant. Binds with high affinity to antithrombin III (AT-III). Other sizes are available. Please inqury us for quote.