Learn More
Selleck Chemical LLC Heparin sodium S1346-1g
SDP
Catalog No. 503111614
Encompass_Preferred
Heparin sodium a sulfated polysaccharide belonging to the family of glycosaminoglycans has numerous important biological activities associated with its interaction with diverse proteins Heparin is used as an anticoagulant primarily through its interaction with AT III by enhancing AT-IIImediated inhibition of blood coagulation factors including thrombin and factor Xa Heparin prevents the induction of autophagy
Catalog No. 50-311-1614 Supplier Selleck Chemical LLC Supplier No. S13461G
Heparin sodium a sulfated polysaccharide belonging to the family of glycosaminoglycans has numerous important biological activities associated with its interaction with diverse proteins Heparin is used as an anticoagulant primarily through its interaction with AT III by enhancing AT-IIImediated inhibition of blood coagulation factors including thrombin and factor Xa Heparin prevents the induction of autophagy