Learn More
Medchemexpress LLC HY-17567A 1g , Heparin (sodium salt) CAS:9041-08-1 Purity:>98%
SDP
Catalog No. 502261009
Encompass_Preferred
HY-17567A 1g Heparin (sodium salt) CAS: 9041-08-1 Heparin sodium salt (Sodium heparin) is an anticoagulant which binds reversibly to antithrombin III (ATIII) and greatly accelerates the rate at which ATIII inactivates coagulation enzymes thrombin factor IIa and factor Xa. Heparin sodium salt significantly inhibits exosome-cell interactions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1009 Supplier Medchemexpress LLC Supplier No. HY17567A1G
HY-17567A 1g Heparin (sodium salt) CAS: 9041-08-1 Heparin sodium salt (Sodium heparin) is an anticoagulant which binds reversibly to antithrombin III (ATIII) and greatly accelerates the rate at which ATIII inactivates coagulation enzymes thrombin factor IIa and factor Xa. Heparin sodium salt significantly inhibits exosome-cell interactions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.