Learn More
Medchemexpress LLC HY-17567A 500mg , Heparin (sodium salt) Sodium heparinate CAS:2608411 Purity:98%
SDP
Catalog No. 501492385
Encompass_Preferred
Medchemexpress, HY-17567A 500mg Heparin (sodium salt) Sodium heparinate CAS:2608411 Formula:(C14H25NO20S3)n.xNa Purity:98% Heparin (sodium salt) is a highly sulfated glycosaminoglycan, that is widely used as an injectable anticoagulant, and has the highest negative charge density of any known biological molecule. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2385 Supplier Medchemexpress LLC Supplier No. HY17567A500MG
Medchemexpress, HY-17567A 500mg Heparin (sodium salt) Sodium heparinate CAS:2608411 Formula:(C14H25NO20S3)n.xNa Purity:98% Heparin (sodium salt) is a highly sulfated glycosaminoglycan, that is widely used as an injectable anticoagulant, and has the highest negative charge density of any known biological molecule. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.