Learn More
Medchemexpress LLC Heparin sodium salt | 9041-08-1 | 15 kDa (for a specific form) | 25MG
SDP
Catalog No. 5000383944
Encompass_Preferred
Heparin sodium salt (Sodium heparin) is an anticoagulant that reversibly binds to antithrombin III (ATIII), significantly accelerating the rate at which ATIII inactivates coagulation enzymes thrombin factor IIa and factor Xa. It also notably inhibits exosome-cell interactions. Heparin is a complex linear polymer composed of chains of varying lengths and sequences.
  • Acts as a potent anticoagulant.
  • Enhances antithrombin-mediated inhibition of blood coagulation factors.
  • Interacts with FGF-2 and FGFR-1, stabilizing FGF-FGFR binding.
  • Promotes FGFR dimerization.
Catalog No. 50-003-83944 Supplier Medchemexpress LLC Supplier No. 25MGHY17567
Heparin sodium salt (Sodium heparin) is an anticoagulant that reversibly binds to antithrombin III (ATIII), significantly accelerating the rate at which ATIII inactivates coagulation enzymes thrombin factor IIa and factor Xa. It also notably inhibits exosome-cell interactions. Heparin is a complex linear polymer composed of chains of varying lengths and sequences.
  • Acts as a potent anticoagulant.
  • Enhances antithrombin-mediated inhibition of blood coagulation factors.
  • Interacts with FGF-2 and FGFR-1, stabilizing FGF-FGFR binding.
  • Promotes FGFR dimerization.