Learn More
Sigma Aldrich Fine Chemicals Biosciences Heparin sodium salt from porcine intestinal mucosa, 9041-08-1, MFCD00081689, 100ku
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H3149100KU
Encompass
Heparin sodium salt from porcine intestinal mucosa Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture, 100000units. Heparin is mainly responsible for the delay in the coagulation of blood. It enhances the antithrombin-mediated inactivation of proteases in the coagulation pathway.
Catalog No. 50-165-7296