Learn More
Sigma Aldrich Fine Chemicals Biosciences Heparin sodium salt from porcine intestinal mucosa, 9041-08-1, MFCD00081689, 10ku
Catalog No. 501657295
Encompass
Heparin sodium salt from porcine intestinal mucosa Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture, 10000units. Heparin is mainly responsible for the delay in the coagulation of blood. It enhances the antithrombin-mediated inactivation of proteases in the coagulation pathway.
Catalog No. 50-165-7295 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H314910KU
Heparin sodium salt from porcine intestinal mucosa Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture, 10000units. Heparin is mainly responsible for the delay in the coagulation of blood. It enhances the antithrombin-mediated inactivation of proteases in the coagulation pathway.