Learn More
Medchemexpress LLC Heparin sodium salt (MW 15kDa) | 9041-08-1 | 15000 | 500 MG
SDP
Catalog No. 5000387195
Encompass_Preferred
Heparin sodium salt (MW 15kDa) is a polymer of Heparin with a molecular weight of 15kDa. It is an anticoagulant that binds reversibly to antithrombin III (ATIII) and significantly accelerates the rate at which ATIII inactivates coagulation enzymes thrombin factor IIa and factor Xa.
  • Polymer of Heparin with a molecular weight of 15kDa
  • Anticoagulant
  • Binds reversibly to antithrombin III (ATIII)
  • Appears as a white to off-white solid
  • For research use only
  • Greatly accelerates the rate at which ATIII inactivates coagulation enzymes thrombin factor IIa and factor Xa
  • Shows angiogenic response (an increase in vascularization) in late Chorioallantoic Membrane (CAM) assay at 0-150 μM for 72 hours
Catalog No. 50-003-87195 Supplier Medchemexpress LLC Supplier No. HY17567C500MG
Heparin sodium salt (MW 15kDa) is a polymer of Heparin with a molecular weight of 15kDa. It is an anticoagulant that binds reversibly to antithrombin III (ATIII) and significantly accelerates the rate at which ATIII inactivates coagulation enzymes thrombin factor IIa and factor Xa.
  • Polymer of Heparin with a molecular weight of 15kDa
  • Anticoagulant
  • Binds reversibly to antithrombin III (ATIII)
  • Appears as a white to off-white solid
  • For research use only
  • Greatly accelerates the rate at which ATIII inactivates coagulation enzymes thrombin factor IIa and factor Xa
  • Shows angiogenic response (an increase in vascularization) in late Chorioallantoic Membrane (CAM) assay at 0-150 μM for 72 hours