Learn More
Medchemexpress LLC Heparin sodium salt (MW 15kDa) | 9041-08-1 | 15000 (Average) | 1 G
SDP
Catalog No. 5000387337
Encompass_Preferred
Heparin sodium salt (MW 15kDa) is a polymer of Heparin with an average molecular weight of 15kDa. It functions as an anticoagulant by reversibly binding to antithrombin III (ATIII), significantly accelerating ATIII's rate of inactivating coagulation enzymes thrombin factor IIa and factor Xa.
  • Induces an angiogenic response (increased vascularization) in a late Chorioallantoic Membrane (CAM) assay.
Catalog No. 50-003-87337 Supplier Medchemexpress LLC Supplier No. HY17567C1G
Heparin sodium salt (MW 15kDa) is a polymer of Heparin with an average molecular weight of 15kDa. It functions as an anticoagulant by reversibly binding to antithrombin III (ATIII), significantly accelerating ATIII's rate of inactivating coagulation enzymes thrombin factor IIa and factor Xa.
  • Induces an angiogenic response (increased vascularization) in a late Chorioallantoic Membrane (CAM) assay.