Learn More
Medchemexpress LLC Heparin sodium salt (MW 15kDa) | 9041-08-1 | 15000 (Average) | 5 G
SDP
Catalog No. 5000387389
Encompass_Preferred
Heparin sodium salt (MW 15kDa) is a polymer of Heparin with a molecular weight of 15kDa. It functions as an anticoagulant by reversibly binding to antithrombin III (ATIII), which significantly increases the rate at which ATIII inactivates coagulation enzymes thrombin factor IIa and factor Xa. This product has shown angiogenic response in late Chorioallantoic Membrane (CAM) assay. It is suitable for laboratory centrifugation and storage.
  • Acts as an anticoagulant
  • Binds reversibly to antithrombin III (ATIII)
  • Accelerates inactivation of coagulation enzymes
  • Used in various biological activities targeting thrombin and factor Xa
  • Shows angiogenic response in CAM assay
Catalog No. 50-003-87389 Supplier Medchemexpress LLC Supplier No. HY17567C5G
Heparin sodium salt (MW 15kDa) is a polymer of Heparin with a molecular weight of 15kDa. It functions as an anticoagulant by reversibly binding to antithrombin III (ATIII), which significantly increases the rate at which ATIII inactivates coagulation enzymes thrombin factor IIa and factor Xa. This product has shown angiogenic response in late Chorioallantoic Membrane (CAM) assay. It is suitable for laboratory centrifugation and storage.
  • Acts as an anticoagulant
  • Binds reversibly to antithrombin III (ATIII)
  • Accelerates inactivation of coagulation enzymes
  • Used in various biological activities targeting thrombin and factor Xa
  • Shows angiogenic response in CAM assay