Learn More
Sigma Aldrich Fine Chemicals Biosciences Heparinase I and III Blend
Catalog No. 502904960
Encompass
Heparinase is an inducible non-extracellular heparin-degrading enzyme. Three types of heparinises are produced by flavobacterium heparinum and contains specific sequences of heparin.
Catalog No. 50-290-4960 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H391750UN
Heparinase is an inducible non-extracellular heparin-degrading enzyme. Three types of heparinises are produced by flavobacterium heparinum and contains specific sequences of heparin.