Learn More
Sigma Aldrich Fine Chemicals Biosciences Heparinase I and III Blend
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H3917100UN
Encompass
Heparinase is an inducible non-extracellular heparin-degrading enzyme. Three types of heparinises are produced by flavobacterium heparinum and contains specific sequences of heparin.
Catalog No. 50-290-4710