Learn More
Medchemexpress LLC Heparinase II P. he 500ug | 500ug
SDP
Catalog No. 5000194696
Encompass_Preferred
Heparinase II protein is an enzyme that cleaves heparin and heparan sulfate glycosaminoglycans using a beta elimination mechanism Specifically it cleaves heparin at the -D-GlcNp2S6S(1- 4) -L-IdoAp2S site and at the -D-GlcNp2Ac(or 2S)6OH(1- 4)beta-D-GlcAp position Site of cleavage of heparan sulfate Heparinase II Protein P heparinus (His) is the recombinant Heparinase II protein expressed by E coli with N-His labeled tag
Catalog No. 50-001-94696 Supplier Medchemexpress LLC Supplier No. HYP79221500UG
Heparinase II protein is an enzyme that cleaves heparin and heparan sulfate glycosaminoglycans using a beta elimination mechanism Specifically it cleaves heparin at the -D-GlcNp2S6S(1- 4) -L-IdoAp2S site and at the -D-GlcNp2Ac(or 2S)6OH(1- 4)beta-D-GlcAp position Site of cleavage of heparan sulfate Heparinase II Protein P heparinus (His) is the recombinant Heparinase II protein expressed by E coli with N-His labeled tag