Learn More
Medchemexpress LLC Heparinase III P. h 10ug | 10ug
SDP
Catalog No. 5000194589
Encompass_Preferred
Heparinase III Protein is the unique member of the heparinase family of heparin-degrading lyases that recognizes the ubiquitous cell-surface heparan sulfate proteoglycans as its primary substrate Heparinase III functions in cleaving metazoan heparan sulfate and providing carbon nitrogen and sulfate sources for microorganisms Furthermore Heparinase III is cardioprotective in a dose-dependent manner preserves endothelial function and attenuates PMN adherence to the coronary vascular endothelium Heparinase III Protein P heparinus (His) is the recombinant Heparinase III protein expressed by E coli with N-6 His labeled tag
Catalog No. 50-001-94589 Supplier Medchemexpress LLC Supplier No. HYP7921110UG
Heparinase III Protein is the unique member of the heparinase family of heparin-degrading lyases that recognizes the ubiquitous cell-surface heparan sulfate proteoglycans as its primary substrate Heparinase III functions in cleaving metazoan heparan sulfate and providing carbon nitrogen and sulfate sources for microorganisms Furthermore Heparinase III is cardioprotective in a dose-dependent manner preserves endothelial function and attenuates PMN adherence to the coronary vascular endothelium Heparinase III Protein P heparinus (His) is the recombinant Heparinase III protein expressed by E coli with N-6 His labeled tag