Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Heparitin sulfate lyase | 37290-86-1 | 10 U
SDP

Catalog No. 5000399432
Encompass_Preferred

Heparitin sulfate lyase (Heparinase III) is a glycosidic lyase targeting heparan sulfate and heparin. This enzyme is considered promising for research related to low-molecular-weight heparin production and cancers.

  • Solid appearance
  • White to off-white color
  • EC number: 4.2.2.8
  • Unit defined as the amount of enzyme that forms 0.1 µM hyaluronic acid per hour at pH 7.5 and 30°C
  • Ships with dry ice
  • Store at -80°C for 2 years

Catalog No. 50-003-99432 Supplier Medchemexpress LLC Supplier No. HYP295310U
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heparitin sulfate lyase (Heparinase III) is a glycosidic lyase targeting heparan sulfate and heparin. This enzyme is considered promising for research related to low-molecular-weight heparin production and cancers.

  • Solid appearance
  • White to off-white color
  • EC number: 4.2.2.8
  • Unit defined as the amount of enzyme that forms 0.1 µM hyaluronic acid per hour at pH 7.5 and 30°C
  • Ships with dry ice
  • Store at -80°C for 2 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.