Learn More
Medchemexpress LLC Heparitin sulfate lyase (Heparinase III) | 37290-86-1 | ≥100 U/mg protein | 5 U
SDP
Catalog No. 5000399431
Encompass_Preferred
Heparitin sulfate lyase (Heparinase III) is a glycosidic lyase that targets heparan sulfate and heparin. This enzyme is promising for research related to low-molecular-weight heparin production and various cancers.
  • Glycosidic lyase targeting heparan sulfate and heparin
  • Useful in research for low-molecular-weight heparin production
  • Applicable in cancer research
  • Specific activity: ≥100 U/mg protein
Catalog No. 50-003-99431 Supplier Medchemexpress LLC Supplier No. HYP29535U
Heparitin sulfate lyase (Heparinase III) is a glycosidic lyase that targets heparan sulfate and heparin. This enzyme is promising for research related to low-molecular-weight heparin production and various cancers.
  • Glycosidic lyase targeting heparan sulfate and heparin
  • Useful in research for low-molecular-weight heparin production
  • Applicable in cancer research
  • Specific activity: ≥100 U/mg protein