Learn More
StatLab Hepatitis B MasterTech Special Stain Kit
SDP
Catalog No. 22000051
Change view
Click to view available options
100 mL
Catalog No.
22-000-051 100 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-000-051 Supplier StatLab Supplier No. KTHEP
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Stains for Hepatitis B surface antigen (HBsAg) in two hours and thirty minutes
  • Orcein Stain serves as the primary stain, differentiated to produce red to brown aggregates in the cytoplasm of hepatocytes and Kupffer cells against a yellowish brown background
  • 100 mL Stain Kit yields between 53 and 66 slides
  • A MasterTech Special Stain Kit
Specifications
Product Type Stain Kit