Learn More
Bioworld Hepatitis B Surface Antigen ayw Subtype, Saccharomyces (Recombinant), 50 μg
Supplier:  Bioworld 220601012
Encompass_Preferred
Recombinant Hepatitis B Surface Antigen, ayw subtype (HBsAg-ayw) Source: Saccharomyces cerevisae Physical appearance: Sterile Filtered colourless liquid formulation Formulation: Sterile Filtered solution containing 50% glycerol, 7.4mM phosphate buffer pH-7.2 and 75mM sodium chloride Stability: Recombinant Hepatitis B Surface Antigen should be stored at 4°C between 3-4 weeks and for future use should be stored desiccated below -18°C . Purity: Greater than 95.0% as determined by SDS-PAGE. Applications: 1. Immunochromatography (capture and conjugate). 2. Immunogen for monoclonal antibody production. Usage: BioWORLD's products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-202