Learn More
Bioworld HEPATITIS C VIRUS NS3 GENOT
Supplier:  Bioworld 220601161
Encompass_Preferred
HEPATITIS C VIRUS NS3 GENOTYPE-5(1192-1459), (RECOMBINANT): Recombinant Hepatitis C Virus NS3 Genotype 5 (1192-1459 a.a.) Source:E.coli Background: HCV is a small 50nm, enveloped, single-stranded, positive sense RNAvirus in the family ...
Catalog No. 50-253-217
 Restricted Item