Learn More
Bioworld HEPATITIS C VIRUS NS3 GENOT
Supplier:  Bioworld 220601091
Encompass_Preferred
HEPATITIS C VIRUS NS3 GENOTYPE-2B(1192-1459), (RECOMBINANT): Recombinant Hepatitis C Virus nonstructural protein NS3 Subtype 2b (1192-1459) Source:E.coli Background: HCV is a small 50nm, enveloped, single-stranded, positive sense RNA...
Catalog No. 50-253-210
 Restricted Item