Learn More
Bioworld HEPATITIS C VIRUS NS3 GENOT
Supplier:  Bioworld 220601111
Encompass_Preferred
HEPATITIS C VIRUS NS3 GENOTYPE-2C (1192-1459), (RECOMBINANT): Recombinant Hepatitis C Virus nonstructural protein NS3 Subtype 2c(1192-1459) Source:E.coli Background: HCV is a small 50nm, enveloped, single-stranded, positive sense RNA...
Catalog No. 50-253-212