Learn More
Bioworld HEPATITIS C VIRUS NS3 GENOT
Supplier:  Bioworld 220601491
Encompass_Preferred
HEPATITIS C VIRUS NS3 GENOTYPE-1C, (RECOMBINANT): Recombinant Hepatitis C Virus nonstructural protein NS3 Subtype 1c (1192-1459) Source:E.coli Background: HCV is a small 50nm, enveloped, single-stranded, positive sense RNAvirus in th...
Catalog No. 50-253-250
 Restricted Item