Learn More
Bioworld HEPATITIS C VIRUS NS4 A+B
Supplier:  Bioworld 220601551
Encompass_Preferred
Hepatitis C Virus NS4 a+b, Biotin Labeled (Recombinant) Source:E.coli Background: HCV is a small 50nm, enveloped, single-stranded, positive sense RNAvirus in the family Flaviviridae. HCV has a high rate of replication with approximate...
Catalog No. 50-253-256