Learn More
Bioworld HEPATITIS C VIRUS NS5 BIOT
Supplier:  Bioworld 220601631
Encompass_Preferred
Hepatitis C Virus NS5, Biotin Labeled (Recombinant) Source:E.coli Background: HCV is a small 50nm, enveloped, single-stranded, positive sense RNAvirus in the family Flaviviridae. HCV has a high rate of replication with approximately o...
Catalog No. 50-253-264