Learn More
Bioworld HEPATITIS C VIRUS NS5GENOTY
Supplier:  Bioworld 220601591
Encompass_Preferred
HEPATITIS C VIRUS NS5GENOTYPE-1(RECOMBINANT): Hepatitis C Virus NS5 Genotype-1 (Recombinant) Source:E.coli Background: HCV is a small 50nm, enveloped, single-stranded, positive sense RNAvirus in the family Flaviviridae. HCV has a hi...
Catalog No. 50-253-260