Learn More
Bioworld HEPATITIS C VIRUS NS5GENOTY
Supplier:  Bioworld 220601221
Encompass_Preferred
HEPATITIS C VIRUS NS5GENOTYPE-1B (RECOMBINANT): Hepatitis C Virus NS5 Genotype-1b (Recombinant) Source:E.coli Background: HCV is a small 50nm, enveloped, single-stranded, positive sense RNAvirus in the family Flaviviridae. HCV has a...
Catalog No. 50-253-223