Learn More
Bioworld HEPATITIS C VIRUS NUCLEOCAP
Supplier:  Bioworld 220601351
Encompass_Preferred
HEPATITIS C VIRUS NUCLEOCAPSID (CORE) GENOTYPE-1A, (RECOMBINANT): Recombinant Hepatitis C Virus Nucleocapsid Genotype 1a Source:E.coli Background: The hepatitis C virus (HCV) core protein represents the first 191 amino acids of the v...
Catalog No. 50-253-236