Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Hepatitis E Mixed Titer Panel
SDP

Catalog No. 22157292 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
24 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-292 24 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-157-292 Supplier ZeptoMetrix Supplier No. KZMC003
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For quantity control

Specifications

Content And Storage -20°C
Product Type Hepatitis E Mixed Titer Panel
Quantity 24 x 0.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.