Learn More
Carbosynth LLC. Purified recombinant Hepatitis E Virus protein (HRP)
SDP
Supplier:  Carbosynth LLC. 65IH351ML
Encompass_Preferred
Hepatitis E virus (HEV) is a positive-sense single-stranded RNA icosahedral virus with a 7.5 kilobase genome. HEV has a fecal-oral transmission route. It is one of five known hepatitis viruses: A, B, C, D, and E.
Catalog No. NC1608110